Assay Details
| Gene Symbol: OCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850585.1 | XP_028706418.1 | 3460 | 62 | ||
| GenBank mRNA | JV716022.1 | - | 723 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OCA2| Gene Symbol: OCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850585.1 | XP_028706418.1 | 3460 | 62 | ||
| GenBank mRNA | JV716022.1 | - | 723 | 62 | ||
Set Membership: |
|