Assay Details
| Gene Symbol: FA2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194422.3 | NP_001181351.2 | 865 | 62 | ||
| GenBank mRNA | JU336356.1 | - | 846 | 62 | ||
| JV723233.1 | - | 772 | 62 | |||
More Information
Set Membership: |
|

FA2H
Genomic Map