Assay Details
| Gene Symbol: RAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261638.1 | NP_001248567.1 | 715 | 77 | ||
| GenBank mRNA | DQ148142.1 | - | 322 | 77 | ||
| JU335752.1 | - | 697 | 77 | |||
| JU471889.1 | - | 701 | 77 | |||
| JV045701.1 | - | 715 | 77 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAP1A
Genomic Map