Assay Details
| Gene Symbol: SLC35E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133065.2 | XP_014988551.2 | 1461 | 70 | ||
| XM_028841392.1 | XP_028697225.1 | 1464 | 70 | |||
| XM_028841394.1 | XP_028697227.1 | 1461 | 70 | |||
| GenBank mRNA | JU336202.1 | - | 863 | 70 | ||
| JU470982.1 | - | 598 | 70 | |||
| JV044391.1 | - | 674 | 70 | |||
| JV564969.1 | - | 834 | 70 | |||
| JV627632.1 | - | 887 | 70 | |||
| JV667655.1 | - | 928 | 70 | |||
More Information
Set Membership: |
|

SLC35E2
Genomic Map