Assay Details
| Gene Symbol: MSRB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257851.1 | NP_001244780.1 | 463 | 75 | ||
| GenBank mRNA | JU333783.1 | - | 463 | 75 | ||
| JU333784.1 | - | 463 | 75 | |||
| JU333785.1 | - | 463 | 75 | |||
| JU333786.1 | - | 463 | 75 | |||
| JU333787.1 | - | 463 | 75 | |||
| JU333788.1 | - | 463 | 75 | |||
| JU333789.1 | - | 463 | 75 | |||
| JU333790.1 | - | 463 | 75 | |||
| JU333791.1 | - | 463 | 75 | |||
| JU333792.1 | - | 463 | 75 | |||
| JU471349.1 | - | 447 | 75 | |||
| JU475027.1 | - | 527 | 75 | |||
| JV046648.1 | - | 592 | 75 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MSRB3
Genomic Map