Assay Details
| Gene Symbol: LARGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260905.1 | NP_001247834.1 | 1732 | 60 | ||
| XM_015150010.2 | XP_015005496.1 | 1871 | 60 | |||
| XM_015150011.2 | XP_015005497.1 | 1783 | 60 | |||
| XM_028827420.1 | XP_028683253.1 | 1538 | 60 | |||
| XM_028827421.1 | XP_028683254.1 | 1408 | 60 | |||
| XM_028827422.1 | XP_028683255.1 | 1395 | 60 | |||
| GenBank mRNA | JU337091.1 | - | 1732 | 60 | ||
| JV701220.1 | - | 1803 | 60 | |||
More Information
Set Membership: |
|

LARGE1
Genomic Map