Assay Details
| Gene Symbol: NTMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266620.1 | NP_001253549.1 | 355 | 65 | ||
| XM_015116599.2 | XP_014972085.1 | 404 | 65 | |||
| XM_028834395.1 | XP_028690228.1 | 440 | 65 | |||
| GenBank mRNA | JU319952.1 | - | 331 | 65 | ||
| JU473314.1 | - | 297 | 65 | |||
| JV045521.1 | - | 355 | 65 | |||
| JV490594.1 | - | 241 | 65 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NTMT1
Genomic Map