Assay Details
| Gene Symbol: SESN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261068.1 | NP_001247997.1 | 360 | 63 | ||
| XM_028845738.1 | XP_028701571.1 | 364 | 63 | |||
| GenBank mRNA | JU477431.1 | - | 360 | 63 | ||
| JV048393.1 | - | 381 | 63 | |||
| JV635748.1 | - | 380 | 63 | |||
More Information
Set Membership: |
|

SESN2
Genomic Map