Assay Details
| Gene Symbol: EFNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265683.2 | NP_001252612.1 | 198 | 98 | ||
| XM_015109938.1 | XP_014965424.1 | 237 | 98 | |||
| GenBank mRNA | JV667357.1 | - | 198 | 98 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EFNA3
Genomic Map