Assay Details
| Gene Symbol: KLHL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261738.1 | NP_001248667.1 | 1684 | 72 | ||
| XM_015136953.2 | XP_014992439.1 | 1963 | 72 | |||
| XM_015136955.2 | XP_014992441.1 | 7070 | 72 | |||
| XM_028846771.1 | XP_028702604.1 | 1797 | 72 | |||
| XM_028846772.1 | XP_028702605.1 | 1742 | 72 | |||
| XM_028846773.1 | XP_028702606.1 | 2779 | 72 | |||
| XM_028846774.1 | XP_028702607.1 | 1951 | 72 | |||
| GenBank mRNA | JU321841.1 | - | 1684 | 72 | ||
More Information
Set Membership: |
|

KLHL32
Genomic Map