Assay Details
| Gene Symbol: CORO1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265934.1 | NP_001252863.1 | 856 | 62 | ||
| XM_015152713.2 | XP_015008199.1 | 1029 | 62 | |||
| XM_028828899.1 | XP_028684732.1 | 1312 | 62 | |||
| GenBank mRNA | JU320741.1 | - | 781 | 62 | ||
| JU471222.1 | - | 856 | 62 | |||
| JV452993.1 | - | 783 | 62 | |||
More Information
Set Membership: |
|

CORO1C
Genomic Map