Assay Details
| Gene Symbol: SMPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266860.1 | NP_001253789.1 | 493 | 67 | ||
| XM_028846674.1 | XP_028702507.1 | 290 | 67 | |||
| GenBank mRNA | JU473217.1 | - | 547 | 67 | ||
| JV043467.1 | - | 493 | 67 | |||
More Information
Set Membership: |
|

SMPD2
Genomic Map