Assay Details
| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002799787.3 | XP_002799833.2 | 2083 | 77 | ||
| GenBank mRNA | JU327584.1 | - | 1375 | 77 | ||
| JV713175.1 | - | 1517 | 77 | |||
More Information
Set Membership: |
|

GUCY1A2
Genomic Map