Assay Details
| Gene Symbol: WDR83OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194406.1 | NP_001181335.1 | 408 | 69 | ||
| GenBank mRNA | JU320875.1 | - | 317 | 69 | ||
| JU470489.1 | - | 252 | 69 | |||
| JV043684.1 | - | 192 | 69 | |||
| JV566224.1 | - | 409 | 69 | |||
| JV639485.1 | - | 361 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

WDR83OS
Genomic Map