Assay Details
| Gene Symbol: GPR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266567.1 | NP_001253496.1 | 172 | 157 | ||
| XM_015127183.2 | XP_014982669.1 | 223 | 157 | |||
| GenBank mRNA | JU321287.1 | - | 172 | 157 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266567.1, XM_015127183.2 transcripts.
Set Membership: |
|

GPR34
Genomic Map