Assay Details
| Gene Symbol: NPTXR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150374.2 | XP_015005860.2 | 1467 | 72 | ||
| GenBank mRNA | JU333695.1 | - | 1307 | 72 | ||
| JU333696.1 | - | 1307 | 72 | |||
| JV500724.1 | - | 931 | 72 | |||
| JV611227.1 | - | 1430 | 72 | |||
More Information
Set Membership: |
|

NPTXR
Genomic Map