Assay Details
| Gene Symbol: FGF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194006.3 | NP_001180935.2 | 1602 | 61 | ||
| XM_015132497.2 | XP_014987983.2 | 410 | 61 | |||
| XM_028843240.1 | XP_028699073.1 | 554 | 61 | |||
| XM_028843241.1 | XP_028699074.1 | 551 | 61 | |||
| XM_028843242.1 | XP_028699075.1 | 400 | 61 | |||
| XM_028843243.1 | XP_028699076.1 | 172 | 61 | |||
| GenBank mRNA | JU319790.1 | - | 1110 | 61 | ||
| JU333436.1 | - | 360 | 61 | |||
| JV498493.1 | - | 365 | 61 | |||
| JV565331.1 | - | 1112 | 61 | |||
| JV601740.1 | - | 1127 | 61 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FGF12
Genomic Map