Assay Details
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194329.1 | NP_001181258.1 | 172 | 58 | ||
| XM_015127341.2 | XP_014982827.1 | 208 | 58 | |||
| XM_028841830.1 | XP_028697663.1 | 213 | 58 | |||
| GenBank mRNA | JV555937.1 | - | 47 | 58 | ||
| JV667462.1 | - | 125 | 58 | |||
More Information
Set Membership: |
|

SYP
Genomic Map