Assay Details
| Gene Symbol: UTP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135769.2 | XP_014991255.2 | 85 | |||
| GenBank mRNA | JU334795.1 | - | 462 | 85 | ||
| JU334796.1 | - | 462 | 85 | |||
| JU334797.1 | - | 462 | 85 | |||
| JU334798.1 | - | 462 | 85 | |||
| JU470969.1 | - | 462 | 85 | |||
| JV704081.1 | - | 529 | 85 | |||
More Information
Set Membership: |
|

UTP11
Genomic Map