Assay Details
| Gene Symbol: LPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847001.1 | XP_028702834.1 | 4743 | 57 | ||
| GenBank mRNA | J04635.1 | - | 2829 | 57 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LPA
Genomic Map