Assay Details
| Gene Symbol: E2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102839.4 | XP_001102839.1 | 1001 | 88 | ||
| GenBank mRNA | JU470817.1 | - | 1006 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

E2F2| Gene Symbol: E2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001102839.4 | XP_001102839.1 | 1001 | 88 | ||
| GenBank mRNA | JU470817.1 | - | 1006 | 88 | ||
Set Membership: |
|