Assay Details
| Gene Symbol: LOC722133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150950.2 | XP_015006436.2 | 216 | 106 | ||
| GenBank mRNA | JU336156.1 | - | 67 | 106 | ||
| JV048104.1 | - | 66 | 106 | |||
| JV660182.1 | - | 74 | 106 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015150950.2 transcripts.
Set Membership: |
|

LOC722133
Genomic Map