Assay Details
| Gene Symbol: SOCS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095656.4 | XP_001095656.2 | 107 | |||
| GenBank mRNA | JU321288.1 | - | 77 | 107 | ||
| JV727046.1 | - | 63 | 107 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001095656.4 transcripts.
Set Membership: |
|

SOCS6
Genomic Map