Assay Details
Gene Symbol: NDST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266766.2 | NP_001253695.1 | 2897 | 66 | ||
XM_015141188.2 | XP_014996674.1 | 3045 | 66 | |||
XM_015141189.2 | XP_014996675.1 | 3025 | 66 | |||
XM_015141190.2 | XP_014996676.1 | 2730 | 66 | |||
XM_015141192.2 | XP_014996678.1 | 2220 | 66 | |||
XM_015141193.2 | XP_014996679.1 | 2200 | 66 | |||
XM_015141194.2 | XP_014996680.1 | 2220 | 66 | |||
XM_028849261.1 | XP_028705094.1 | 2908 | 66 | |||
XM_028849262.1 | XP_028705095.1 | 2091 | 66 | |||
GenBank mRNA | JU319746.1 | - | 2894 | 66 | ||
JU471199.1 | - | 2905 | 66 | |||
JV046942.1 | - | 2902 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
