Assay Details
| Gene Symbol: CERK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266852.1 | NP_001253781.1 | 309 | 68 | ||
| XM_028827371.1 | XP_028683204.1 | 651 | 68 | |||
| GenBank mRNA | JU320988.1 | - | 298 | 68 | ||
| JU320989.1 | - | 298 | 68 | |||
| JU471027.1 | - | 309 | 68 | |||
| JV590989.1 | - | 68 | ||||
| JV668404.1 | - | 311 | 68 | |||
More Information
Set Membership: |
|

CERK
Genomic Map