Assay Details
| Gene Symbol: SLC7A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257753.1 | NP_001244682.1 | 1686 | 70 | ||
| XM_015143288.2 | XP_014998774.1 | 1658 | 70 | |||
| XM_015143289.2 | XP_014998775.1 | 1748 | 70 | |||
| GenBank mRNA | JU321297.1 | - | 1686 | 70 | ||
| JV045047.1 | - | 1697 | 70 | |||
| JV703840.1 | - | 2189 | 70 | |||
More Information
Set Membership: |
|

SLC7A8
Genomic Map