Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110764.2 | XP_014966250.1 | 1792 | 115 | ||
| XM_015110771.2 | XP_014966257.1 | 1792 | 115 | |||
| GenBank mRNA | JV635954.1 | - | 900 | 115 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map