Assay Details
| Gene Symbol: EZR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137038.2 | XP_014992524.1 | 229 | 80 | ||
| GenBank mRNA | JU336246.1 | - | 220 | 80 | ||
| JU470688.1 | - | 154 | 80 | |||
| JU470689.1 | - | 154 | 80 | |||
More Information
Set Membership: |
|

EZR
Genomic Map