Assay Details
| Gene Symbol: LOC114675624 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842839.1 | XP_028698672.1 | 430 | 143 | ||
| Gene Symbol: CASK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261010.1 | NP_001247939.1 | 1609 | 143 | ||
| XM_015127187.2 | XP_014982673.1 | 1758 | 143 | |||
| XM_015127188.2 | XP_014982674.1 | 1759 | 143 | |||
| XM_015127189.2 | XP_014982675.1 | 1740 | 143 | |||
| XM_015127190.2 | XP_014982676.1 | 1740 | 143 | |||
| XM_015127191.2 | XP_014982677.1 | 1606 | 143 | |||
| XM_015127192.2 | XP_014982678.1 | 1721 | 143 | |||
| XM_015127193.2 | XP_014982679.1 | 1758 | 143 | |||
| XM_015127194.2 | XP_014982680.1 | 1751 | 143 | |||
| XM_015127195.2 | XP_014982681.1 | 1740 | 143 | |||
| XM_015127196.2 | XP_014982682.1 | 1598 | 143 | |||
| XM_015127197.2 | XP_014982683.1 | 1758 | 143 | |||
| XM_015127198.2 | XP_014982684.1 | 1721 | 143 | |||
| XM_028841793.1 | XP_028697626.1 | 1752 | 143 | |||
| GenBank mRNA | JU320476.1 | - | 1609 | 143 | ||
| JU328554.1 | - | 598 | 143 | |||
| JU336863.1 | - | 1609 | 143 | |||
More Information
Set Membership: |
|

LOC114675624
Genomic Map