Assay Details
| Gene Symbol: AP1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845615.1 | XP_028701448.1 | 341 | 61 | ||
| GenBank mRNA | JU332851.1 | - | 294 | 61 | ||
| JV045643.1 | - | 287 | 61 | |||
| JV451788.1 | - | 248 | 61 | |||
| JV567541.1 | - | 300 | 61 | |||
| JV669583.1 | - | 220 | 61 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP1S1
Genomic Map