Assay Details
| Gene Symbol: CDK2AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100820.3 | XP_001100820.1 | 757 | 66 | ||
| XM_015153071.2 | XP_015008557.1 | 544 | 66 | |||
| XM_015153072.2 | XP_015008558.1 | 347 | 66 | |||
| XM_015153073.2 | XP_015008559.1 | 335 | 66 | |||
| XM_028830085.1 | XP_028685918.1 | 800 | 66 | |||
| XM_028830086.1 | XP_028685919.1 | 455 | 66 | |||
| GenBank mRNA | JU349014.1 | - | 245 | 66 | ||
| JU477339.1 | - | 361 | 66 | |||
| JV048329.1 | - | 343 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDK2AP1
Genomic Map