Assay Details
| Gene Symbol: KAZALD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109857.4 | XP_001109857.1 | 1595 | 118 | ||
| XM_028826786.1 | XP_028682619.1 | 837 | 118 | |||
| XM_028826787.1 | XP_028682620.1 | 879 | 118 | |||
| XM_028826788.1 | XP_028682621.1 | 1242 | 118 | |||
More Information
Set Membership: |
|

KAZALD1
Genomic Map