Assay Details
| Gene Symbol: RGS7BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266063.1 | NP_001252992.1 | 171 | 75 | ||
| XM_015140026.2 | XP_014995512.1 | 655 | 75 | |||
| GenBank mRNA | JU334488.1 | - | 171 | 75 | ||
| JV683952.1 | - | 173 | 75 | |||
More Information
Set Membership: |
|

RGS7BP
Genomic Map