Assay Details
| Gene Symbol: ZDHHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261328.1 | NP_001248257.1 | 258 | 61 | ||
| XM_015149726.2 | XP_015005212.1 | 327 | 61 | |||
| GenBank mRNA | JU321891.1 | - | 258 | 61 | ||
| JU322060.1 | - | 258 | 61 | |||
| JV566748.1 | - | 283 | 61 | |||
More Information
Set Membership: |
|

ZDHHC8
Genomic Map