Assay Details
| Gene Symbol: TMLHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099674.3 | XP_001099674.2 | 930 | 79 | ||
| XM_015128581.2 | XP_014984067.1 | 588 | 79 | |||
| XM_015128582.2 | XP_014984068.1 | 748 | 79 | |||
| XM_015128583.2 | XP_014984069.1 | 930 | 79 | |||
| XM_028842201.1 | XP_028698034.1 | 1060 | 79 | |||
| XM_028842202.1 | XP_028698035.1 | 930 | 79 | |||
| GenBank mRNA | JU321406.1 | - | 885 | 79 | ||
| JU476234.1 | - | 899 | 79 | |||
More Information
Set Membership: |
|

TMLHE
Genomic Map