Assay Details
| Gene Symbol: RELCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122137.2 | XP_014977623.1 | 2101 | 74 | ||
| XM_015122138.2 | XP_014977624.1 | 2101 | 74 | |||
| XM_015122139.2 | XP_014977625.1 | 2101 | 74 | |||
| XM_015122140.2 | XP_014977626.1 | 2035 | 74 | |||
| XM_015122141.2 | XP_014977627.1 | 2035 | 74 | |||
| XM_015122142.2 | XP_014977628.1 | 2029 | 74 | |||
| XM_015122143.2 | XP_014977629.1 | 1748 | 74 | |||
| XM_015122145.2 | XP_014977631.1 | 2101 | 74 | |||
| XM_028838171.1 | XP_028694004.1 | 2101 | 74 | |||
| XM_028838172.1 | XP_028694005.1 | 2035 | 74 | |||
| XM_028838173.1 | XP_028694006.1 | 459 | 74 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RELCH
Genomic Map