Assay Details
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261191.1 | NP_001248120.1 | 1382 | 63 | ||
XM_015126963.2 | XP_014982449.1 | 1410 | 63 | |||
GenBank mRNA | JU320745.1 | - | 1367 | 63 | ||
JU471630.1 | - | 1382 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
