Assay Details
| Gene Symbol: NAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194588.2 | NP_001181517.1 | 918 | 64 | ||
| GenBank mRNA | JU336919.1 | - | 924 | 64 | ||
| JU477654.1 | - | 929 | 64 | |||
| JV048585.1 | - | 946 | 64 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NAPA
Genomic Map