Assay Details
| Gene Symbol: GRM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261246.1 | NP_001248175.1 | 1349 | 92 | ||
| XM_015134808.2 | XP_014990294.1 | 1741 | 92 | |||
| XM_028844840.1 | XP_028700673.1 | 1953 | 92 | |||
| GenBank mRNA | JU322197.1 | - | 1349 | 92 | ||
More Information
Set Membership: |
|

GRM8
Genomic Map