Assay Details
| Gene Symbol: C15H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260888.1 | NP_001247817.1 | 439 | 71 | ||
| GenBank mRNA | JU322704.1 | - | 572 | 71 | ||
| JU473192.1 | - | 439 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C15H9orf64
Genomic Map