Assay Details
| Gene Symbol: HIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118215.2 | XP_014973701.1 | 1088 | 147 | ||
| GenBank mRNA | JV649743.1 | - | 142 | 147 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIC1| Gene Symbol: HIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118215.2 | XP_014973701.1 | 1088 | 147 | ||
| GenBank mRNA | JV649743.1 | - | 142 | 147 | ||
Set Membership: |
|