Assay Details
| Gene Symbol: ASIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261065.1 | NP_001247994.1 | 1034 | 91 | ||
| XM_015151639.2 | XP_015007125.1 | 2847 | 91 | |||
| GenBank mRNA | JU334515.1 | - | 1034 | 91 | ||
| JV684155.1 | - | 1061 | 91 | |||
More Information
Set Membership: |
|

ASIC1
Genomic Map