Assay Details
| Gene Symbol: RBM38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257471.1 | NP_001244400.1 | 393 | 75 | ||
| XM_015148714.2 | XP_015004200.1 | 639 | 75 | |||
| GenBank mRNA | JU335346.1 | - | 393 | 75 | ||
| JU476300.1 | - | 517 | 75 | |||
| JV047580.1 | - | 536 | 75 | |||
More Information
Set Membership: |
|

RBM38
Genomic Map