Assay Details
| Gene Symbol: POLR2G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266524.1 | NP_001253453.1 | 577 | 63 | ||
| XM_015113755.2 | XP_014969241.1 | 739 | 63 | |||
| GenBank mRNA | JU320820.1 | - | 539 | 63 | ||
| JU473750.1 | - | 512 | 63 | |||
| JV047421.1 | - | 577 | 63 | |||
| JV471922.1 | - | 432 | 63 | |||
More Information
Set Membership: |
|

POLR2G
Genomic Map