Assay Details
| Gene Symbol: CD2AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266309.1 | NP_001253238.1 | 1969 | 81 | ||
| XM_028846750.1 | XP_028702583.1 | 1468 | 81 | |||
| GenBank mRNA | JU323155.1 | - | 1612 | 81 | ||
| JU472085.1 | - | 1969 | 81 | |||
More Information
Set Membership: |
|

CD2AP
Genomic Map