Assay Details
| Gene Symbol: LOC694442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084988.3 | XP_001084988.2 | 2767 | 105 | ||
| XM_001085113.3 | XP_001085113.2 | 3060 | 105 | |||
More Information
Set Membership: |
|

LOC694442
Genomic Map