Assay Details
| Gene Symbol: DR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257839.1 | NP_001244768.1 | 565 | 88 | ||
| GenBank mRNA | JU333253.1 | - | 565 | 88 | ||
| JU470983.1 | - | 1036 | 88 | |||
| JU470984.1 | - | 1030 | 88 | |||
| JV045542.1 | - | 832 | 88 | |||
| JV637694.1 | - | 1102 | 88 | |||
More Information
Set Membership: |
|

DR1
Genomic Map