Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266046.1 | NP_001252975.1 | 1406 | 73 | ||
| XM_028849077.1 | XP_028704910.1 | 1055 | 73 | |||
| GenBank mRNA | JU476525.1 | - | 1406 | 73 | ||
More Information
Set Membership: |
|

PTGER4
Genomic Map