Assay Details
| Gene Symbol: APOO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257632.1 | NP_001244561.1 | 470 | 63 | ||
| XM_015127089.2 | XP_014982575.1 | 471 | 63 | |||
| XM_015127090.2 | XP_014982576.1 | 467 | 63 | |||
| XM_028841700.1 | XP_028697533.1 | 467 | 63 | |||
| GenBank mRNA | JU322255.1 | - | 470 | 63 | ||
| JU471982.1 | - | 473 | 63 | |||
| JV043563.1 | - | 246 | 63 | |||
| JV489744.1 | - | 340 | 63 | |||
More Information
Set Membership: |
|

APOO
Genomic Map