Assay Details
| Gene Symbol: TRAF3IP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266136.1 | NP_001253065.1 | 1562 | 101 | ||
| XM_015137255.2 | XP_014992741.2 | 2000 | 101 | |||
| XM_015137256.2 | XP_014992742.1 | 1965 | 101 | |||
| XM_028846655.1 | XP_028702488.1 | 407 | 101 | |||
| XM_028846656.1 | XP_028702489.1 | 616 | 101 | |||
| GenBank mRNA | JU474061.1 | - | 1562 | 101 | ||
More Information
Set Membership: |
|

TRAF3IP2
Genomic Map